missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP1 Antibody (1D10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26558-50ul
This item is not returnable.
View return policy
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human samples. It is validated for ELISA
Specifications
| GLP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1D10 | |
| ELISA | |
| glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
| A synthesized peptide derived from Human GLP1 [UniProt P01275] | |
| 50 μL | |
| Cancer, Signal Transduction, Stem Cell Markers | |
| 2641 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction