missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP1 Antibody (1D10), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 536.55
Specifications
| Antigen | GLP1 |
|---|---|
| Clone | 1D10 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043681
|
Novus Biologicals
NBP3-26558-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043662
|
Novus Biologicals
NBP3-26558-100ul |
100 μL |
€ 568.00 € 536.55 / 100µL Save € 31.45 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human samples. It is validated for ELISASpecifications
| GLP1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
| A synthesized peptide derived from Human GLP1 [UniProt P01275] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1D10 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2641 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title