Learn More
GLT8D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21232-25ul
 View more versions of this product
Product Code. 18653414
  • € 450.00 / 25µL
Description
GLT8D2 Polyclonal antibody specifically detects GLT8D2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLT8D2
Polyclonal
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
EC 2.4.1, EC 2.4.1.-, FLJ31494, GALA4A, glycosyltransferase 8 domain containing 2, glycosyltransferase 8 domain-containing protein 2, gycosyltransferase
This antibody was developed against Recombinant Protein corresponding to amino acids: MGYLDYRKKAIKDLGISPSTCSFNPGVIVANMTEWKHQRITKQLEKWMQKNVEENLYSSSLGGGVATSPMLIVFHGKYSTINPLWHIRHLGWNPDARYSEHFLQEAKLLH
25 μL
Primary
Human
Purified
Immunocytochemistry
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified
RUO
83468
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.