missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLT8D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 450.00 - € 648.00
Specifications
| Antigen | GLT8D2 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653414
|
Novus Biologicals
NBP3-21232-25ul |
25 μL |
€ 450.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653404
|
Novus Biologicals
NBP3-21232-100ul |
100 μL |
€ 648.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLT8D2 Polyclonal antibody specifically detects GLT8D2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| GLT8D2 | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 2.4.1, EC 2.4.1.-, FLJ31494, GALA4A, glycosyltransferase 8 domain containing 2, glycosyltransferase 8 domain-containing protein 2, gycosyltransferase | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MGYLDYRKKAIKDLGISPSTCSFNPGVIVANMTEWKHQRITKQLEKWMQKNVEENLYSSSLGGGVATSPMLIVFHGKYSTINPLWHIRHLGWNPDARYSEHFLQEAKLLH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 83468 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title