missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamate Receptor 6 Antibody (SR1029), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22252-100ul
This item is not returnable.
View return policy
Description
Glutamate Receptor 6 Monoclonal antibody specifically detects Glutamate Receptor 6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Glutamate Receptor 6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1029 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427 | |
| A synthesized peptide derived from human Glutamate Receptor 6 (Uniprot #: Q13002) | |
| 100 μg | |
| Apoptosis, Cell Biology, GPCR, Neuroscience, Signal Transduction | |
| 2898 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction