missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamate Receptor 6 Antibody (SR1029), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Glutamate Receptor 6 |
|---|---|
| Clone | SR1029 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613964
|
Novus Biologicals
NBP3-22252-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615634
|
Novus Biologicals
NBP3-22252-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutamate Receptor 6 Monoclonal antibody specifically detects Glutamate Receptor 6 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Glutamate Receptor 6 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427 | |
| A synthesized peptide derived from human Glutamate Receptor 6 (Uniprot #: Q13002) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1029 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, GPCR, Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2898 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title