missing translation for 'onlineSavingsMsg'
Learn More

Glycine isopropyl ester hydrochloride, 96%

Product Code. 11409380
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409380 5 g 5g
11499370 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409380 Supplier Thermo Scientific Alfa Aesar Supplier No. H50249.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14019-62-6 | C5H12ClNO2 | 153.606 g/mol

Glycine isopropyl ester hydrochloride can react with indol-3-yl-oxo-acetyl chloride to produce [2-(1H-indol-3-yl)-2-oxo-acetylamino]-acetic acid isopropyl ester. This reaction will need reagent Et3N, and the menstruum benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine isopropyl ester hydrochloride can react with indol-3-yl-oxo-acetyl chloride to produce [2-(1H-indol-3-yl)-2-oxo-acetylamino]-acetic acid isopropyl ester. This reaction will need reagent Et3N, and the menstruum benzene.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14019-62-6
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00151826
InChI Key UBKCIXXGQRZHRO-UHFFFAOYSA-N
Synonym isopropyl 2-aminoacetate hydrochloride, glycine isopropyl ester hydrochloride, propan-2-yl 2-aminoacetate hydrochloride, glycine, 1-methylethyl ester, hydrochloride, h-gly-oipr?cl, glycine, 1-methylethylester, hydrochloride 1:1
PubChem CID 23033482
IUPAC Name propan-2-yl 2-aminoacetate;hydrochloride
SMILES CC(C)OC(=O)CN.Cl

Specifications

Linear Formula NH2CH2CO2CH(CH3)2·HCl
Solubility Information Soluble in water
Quantity 5 g
Formula Weight 153.61
Percent Purity 96%
Chemical Name or Material Glycine isopropyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.