Learn More
Glycine n-propyl ester hydrochloride
Product Code. 11419380
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11419380
Brand: Thermo Scientific Alfa Aesar H50250.03
Request Bulk or Custom Format
CAS: 13049-01-9 | C5H12ClNO2 | 153.606 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13049-01-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00151829
InChI Key YDBMNSABUROTEK-UHFFFAOYSA-N
Synonym propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1
PubChem CID 2759254
IUPAC Name propyl 2-aminoacetate;hydrochloride
SMILES CCCOC(=O)CN.Cl
Specifications
Melting Point 78°C to 80°C
Quantity 1 g
Formula Weight 117.15
Chemical Name or Material Glycine n-propyl ester hydrochloride