missing translation for 'onlineSavingsMsg'
Learn More

Glycolaldehyde diethyl acetal, stab. with ca 0.1% sodium carbonate, 98%

Product Code. p-7044411
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362446 2 g 2g
11372446 10 g 10g
11382446 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362446 Supplier Thermo Scientific Alfa Aesar Supplier No. L06282.04
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 621-63-6 | C6H14O3 | 134.175 g/mol

Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acid and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-63-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00051486
InChI Key IKKUKDZKIIIKJK-UHFFFAOYSA-N
Synonym ethanol, 2,2-diethoxy, glycolaldehyde diethyl acetal, 2,2-diethoxy-ethanol, 2,2-diethoxyethan-1-ol, glycolaldehyde, diethyl acetal, 2,2-diethoxy alcohol, glycolaldehyde diethyl acetal, stab. with ca sodium carbonate, ethanol,2-diethoxy, acmc-20a2bj
PubChem CID 12129
IUPAC Name 2,2-diethoxyethanol
SMILES CCOC(CO)OCC

Specifications

Boiling Point 166°C to 167°C
Flash Point 62°C (143°F)
Refractive Index 1.416
Quantity 2 g
Beilstein 773727
Solubility Information Miscible with water,chloroform and ethyl acetate.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Glycolaldehyde diethyl acetal, stabilized with ≈0.1% sodium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.