missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican-3 Rabbit, Clone: GPC3/1534R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 2719-RBM4-P1
This item is not returnable.
View return policy
Specifications
| Glypican-3 | |
| Cytoplasmic | |
| GPC3/1534R | |
| Unconjugated | |
| NP_004475 | |
| Rabbit | |
| 67kDa | |
| Protein A/G Purified | |
| RUO | |
| 2719 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| GPC3 | |
| Recombinant human full-length GPC3 protein | |
| 293T cells or Hepatocellular carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction