missing translation for 'onlineSavingsMsg'
Learn More

Glypican-3 Rabbit, Clone: GPC3/1534R, enQuireBio™

Rabbit Monoclonal Antibody

€ 199.00 - € 478.00

Specifications

Antigen Glypican-3
Clone GPC3/1534R
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15970108
View Documents
EnquireBio
2719-RBM4-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15980108
View Documents
EnquireBio
2719-RBM4-P1
100 μg
ONLINE EXCLUSIVE PRICE
€ 573.00
€ 478.00 / 100µg
Save € 95.00
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Glypican-3
200μg/mL
Monoclonal
Rabbit
10mM PBS with or without 0.05% BSA and 0.05% azide.
2719
Recombinant human full-length GPC3 protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
67kDa
GPC3/1534R
Immunohistochemistry
Unconjugated
RUO
NP_004475
GPC3
IgG κ
Protein A/G Purified
Cytoplasmic
293T cells or Hepatocellular carcinoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.