missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74902-0.5mg
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| IGHA1 | |
| Purified Human IgA | |
| 0.5 mg | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy alpha chains on human IgA | |
| Store at 4°C. Do not freeze. |
| Immunoassay | |
| 4.5 mg/mL | |
| Immunoglobulin A | |
| Goat | |
| Affinity purified | |
| RUO | |
| 3493 | |
| Human | |
| IgA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction