missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
€ 107.00
Specifications
| Antigen | IgA Heavy Chain |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.Specifications
| IgA Heavy Chain | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Immunoglobulin A | |
| IGHA1 | |
| IgA | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy alpha chains on human IgA |
| 4.5 mg/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| 3493 | |
| Purified Human IgA | |
| Secondary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title