Learn More
Gold shot, 0.8-6mm (0.03-0.2in), Premion™, 99.999% (metals basis)
Product Code. 11371129
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. 11371129
Brand: Thermo Scientific Alfa Aesar 011355.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 7440-57-5
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7440-57-5
Quantity 25 g
Percent Purity 99.999%