Learn More
GOLM1 Antibody (9C2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26550-100ul
 View more versions of this product
Product Code. 30043507
  • € 533.00 / 100µL
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GOLM1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA
9C2
Western Blot 1:500-1:5000, Flow Cytometry, ELISA
bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257
A synthesized peptide derived from Human GOLM1 [UniProt Q8NBJ4]
100 μL
Golgi Apparatus Markers, Signal Transduction
51280
Store at -20 to -70C. Avoid freeze-thaw cycles.