missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody (9C2), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | GOLM1 |
|---|---|
| Clone | 9C2 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
30043648
|
Novus Biologicals
NBP3-26550-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043507
|
Novus Biologicals
NBP3-26550-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISATekniske data
| GOLM1 | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| A synthesized peptide derived from Human GOLM1 [UniProt Q8NBJ4] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9C2 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 51280 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel