missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPHN Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502216
This item is not returnable.
View return policy
Description
Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.Specifications
| GPHN | |
| Monoclonal | |
| Unconjugated | |
| GPHN | |
| GEPH, GPH, GPHRYN, HKPX1, MOCODC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10243 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2C2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NQX3 | |
| GPHN | |
| Full length human recombit protein of human GPHN produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction