missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPHN Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502216S
This item is not returnable.
View return policy
Description
GPHN Monoclonal antibody specifically detects GPHN in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| GPHN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UKS6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29763, 475984 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2C2 | |
| Unconjugated | |
| PACSIN3 | |
| 5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein | |
| Full length human recombinant protein of human PACSIN3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction