Learn More
Poeppelmann™ KAPSTO™ GPN 1000 Hexagonal Caps
Provides optimum protection against corrosion. Poeppelmann™ KAPSTO™ GPN 1000 Hexagonal Caps are manufactured from black PE-LD plastic.
Brand: Poeppelmann™ 10010730000_Y
Additional Details : Weight : 0.50000kg
Description
- Protection for nuts and bolts
- Thick-walled design for higher demands
- Cylindrical outer contour prevents application of spanners
Specifications
Protection for nuts and bolts | |
Black | |
100/Pk. | |
23.6 mm |
Hexagonal Cap | |
19 mm | |
LDPE | |
20 mm |