Learn More
Poeppelmann™ KAPSTO™ GPN 1000 Hexagonal Caps
Provides optimum protection against corrosion. Poeppelmann™ KAPSTO™ GPN 1000 Hexagonal Caps are manufactured from black PE-LD plastic.
€ 13.60 - € 101.00
Product Code | Brand | Diameter (Metric) Outside | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
10616655
|
Poeppelmann™
10010780000 |
39.7 mm | 34 mm | 32 mm |
€ 39.80
Pack of 100 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
10695685
|
Poeppelmann™
10008930000 |
16.8 mm | 14 mm | 13 mm |
€ 14.40
Pack of 100 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10378774
|
Poeppelmann™
10010710000 |
18 mm | 14 mm | 14 mm |
€ 14.75
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
10685685
|
Poeppelmann™
10010690000 |
12.9 mm | 12 mm | 10 mm |
€ 14.40
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10677605
|
Poeppelmann™
10010730000 |
24 mm | 20 mm | 19 mm |
€ 17.65
Pack of 100 |
In Stock
Log in to see stock. |
|||
10409874
|
Poeppelmann™
10082670000 |
21.3 mm | 18 mm | 16 mm |
€ 13.60
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
10517394
|
Poeppelmann™
10010730000_Y |
23.6 mm | 20 mm | 19 mm |
€ 24.70
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
10411225
|
Poeppelmann™
10010740000 |
27.5 mm | 22.5 mm | 22 mm |
€ 24.35
Pack of 100 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
10287994
|
Poeppelmann™
10010760000 |
34 mm | 29 mm | 27 mm |
€ 32.30
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
10606445
|
Poeppelmann™
10110810000 |
55.9 mm | 49 mm | 46 mm |
€ 86.55
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
10207764
|
Poeppelmann™
10010810000 |
55.9 mm | 49 mm | 46 mm |
€ 99.10
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10348974
|
Poeppelmann™
10018110000_Y |
67 mm | 57 mm | 55 mm |
€ 101.00
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
10606655
|
Poeppelmann™
10010770000 |
37.4 mm | 31 mm | 30 mm |
€ 47.55
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
10595874
|
Poeppelmann™
10010720000 |
21.3 mm | 18.5 mm | 17 mm |
€ 18.75
Pack of 100 |
In Stock
Log in to see stock. |
|||
10308574
|
Poeppelmann™
10010800000 |
50.1 mm | 44 mm | 41 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Protection for nuts and bolts
- Thick-walled design for higher demands
- Cylindrical outer contour prevents application of spanners
Specifications
Black | |
LDPE | |
Hexagonal Cap | |
100/Pk. |