Learn More
Poeppelmann™ KAPSTO™ GPN 1000 Hexagonal Caps
Provides optimum protection against corrosion. Poeppelmann™ KAPSTO™ GPN 1000 Hexagonal Caps are manufactured from black PE-LD plastic.
Brand:  Poeppelmann™ 10110810000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10606445
  • € 86.55 / Pack of 100
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Protection for nuts and bolts
  • Thick-walled design for higher demands
  • Cylindrical outer contour prevents application of spanners
Specifications
Specifications
Protection for nuts and bolts
Black
100/Pk.
55.9 mm
Hexagonal Cap
46 mm
LDPE
49 mm