Learn More
Poeppelmann™ GPN 200 Series Natural Form A LDPE Cylindrical Protective Cap
Brand: Poeppelmann™ 20007910000
Additional Details : Weight : 0.50000kg
Specifications
7 mm | |
Protective Cap | |
100 Pack | |
91 mm |
For protection of threaded studs, pipe ends and shafts | |
Natural | |
LDPE |