Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure (22.5mm Insertion Depth)
Brand: Poeppelmann™ 60021800000
Additional Details : Weight : 0.50000kg
Specifications
O.D. x H: 235 x 22.5 mm, 22.5 mm Insertion Depth | |
Tapered Closure | |
25 Pack | |
235 mm |
For use as plugs in bores or internal thread, or as caps for external thread | |
Natural | |
LDPE | |
22.5 mm |