Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 60017400000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10165394
  • € 104.00 / Pack of 25
In Stock
to see stock.
Add to basket
Description
Description
  • 23mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
O.D. x H: 184 x 22.5 mm, 23 mm Insertion Depth
Tapered Closure
25 Pack
184 mm
For use as plugs in bores or internal thread, or as caps for external thread
Natural
LDPE
22.5 mm