Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 70004970000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10606825
  • € 27.55 / Pack of 100
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial internal threads, as well as pipe threads
Specifications
Specifications
Screw Plug Closure
7/16 - 20 UNF
Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1