missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR162 Antibody (1040325) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB10844
This item is not returnable.
View return policy
Description
GPR162 Monoclonal antibody specifically detects GPR162 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| GPR162 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| 1040325 | |
| Unconjugated | |
| A-2, FLJ78837, G protein-coupled receptor 162, Gene-rich cluster gene A protein, GRCAgene rich cluster, A, probable G-protein coupled receptor 162 | |
| Partial synthetic peptide made to an extracellular loop of the human GPR162 protein (between amino acids 1-100) [UniProt Q16538] | |
| 0.1 mg | |
| GPCR, Neuroscience | |
| 27239 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction