missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Smooth Surface Folded Filter Qualitative Filter Papers, Grade 4b

Used for routine analyses like clarification and determination of substances

Brand:  Sartorius™ FT-4-309-125

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10347401

  • € 40.30 / Pack of 100
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N < 0.1%)
  • Wet-strengthened
Specifications

Specifications

Folded
22μm/sec.
4 b
0.15 mm
125 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
125 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers