Learn More
Sartorius™ Smooth Surface Folded Filter Qualitative Filter Papers, Grade 4b
Used for routine analyses like clarification and determination of substances
€ 40.30 - € 319.00
Specifications
Format | Folded |
---|---|
Material | Cotton Linters, Refined Pulp |
Filtration Speed | 22μm/sec. |
For Use With (Application) | For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications |
Grade | 4 b |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10123751
|
Sartorius™
FT-4-309-110 |
110 mm |
€ 61.70
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10347401
|
Sartorius™
FT-4-309-125 |
125 mm |
€ 40.30
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10103851
|
Sartorius™
FT-4-309-150 |
150 mm |
€ 40.30
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10499501
|
Sartorius™
FT-4-309-185 |
185 mm |
€ 40.30
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10694512
|
Sartorius™
FT-4-309-240 |
240 mm |
€ 68.70
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10544571
|
Sartorius™
FT-4-309-250 |
250 mm |
€ 68.70
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10276131
|
Sartorius™
FT-4-309-270 |
270 mm |
€ 75.50
Pack of 100 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11776235
|
Sartorius™
FT-4-309-300 |
300 mm |
€ 99.40
Pack of 100 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10036811
|
Sartorius™
FT-4-309-320 |
320 mm |
€ 99.40
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10347641
|
Sartorius™
FT-4-309-385 |
385 mm |
€ 319.00
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10285891
|
Sartorius™
FT-4-309-090 |
90 mm |
€ 127.00
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
- Made of refined pulp and cotton linters with an >95% α-cellulose content
- Ash content between 0.1–0.15% (grade 100/N < 0.1%)
- Wet-strengthened
Specifications
Folded | |
22μm/sec. | |
4 b | |
0.15 mm |
Cotton Linters, Refined Pulp | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
100 Pack | |
Qualitative Filter Paper |