missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 69 K Smooth Surface Filter Discs Qualitative Filter Papers

Used for analytical and technical purposes.

Marque:  Sartorius™ FT-3-326-070

Informations supplémentaires : Weight : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 11710476

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement »


Description

Description

Used for analytical and technical purposes.
Spécification

Spécification

Disc
65μm/sec.
69 K
0.38 mm
70 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
50 Pack
Qualitative Filter Paper
70 mm
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions