missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 912 Universal Test Paper
Used for general analysis.
Brand: Fisherbrand™
Additional Details : Weight : 0.02700kg
Description
- General control of acid or alkaline reactions
Color:
- Litmus Blue: Blue to red
- Litmus neutral: Purple to blue/red
- Litmus red: Red to blue
Applications:
- Rapid determination of pH value in industrial applications
- General analysis in school
- Used in formation
Specifications
Blue | |
Rapid determination of pH value in industrial applications, general analysis in school, formation | |
Blue Litmus Paper |
Box | |
200 Pack |