Learn More
GRB10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41159
 View more versions of this product
Product Code. 18138557
  • € 532.00 / 0.10mg
Description
GRB10 Polyclonal specifically detects GRB10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GRB10
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
GRB10
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human GRB10. The immunogen is located within amino acids 440 - 490 of GRB10. Amino Acid Squence: EEGHAWRKRSTRMNILG
0.1 mg
Cancer
2887
Human, Mouse, Rat
IgG
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1 - 2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 20 ug/ml
Grb-10, GRB10 adapter protein, GRB10 adaptor protein, GRBIR, GRB-IR, growth factor receptor-bound protein 10, Insulin receptor-binding protein Grb-IR, IRBP, KIAA0207, maternally expressed gene 1, MEG1, RSS
Rabbit
Affinity Purified
RUO
Primary
At least three isoforms of GRB10 are known to exist; this antibody will detect all three isoforms.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.