missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 189.00 - € 532.00
Specifications
| Antigen | GRB10 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685306
|
Novus Biologicals
NBP2-41159-0.025mg |
0.025 mg |
€ 189.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18138557
|
Novus Biologicals
NBP2-41159 |
0.1 mg |
€ 532.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRB10 Polyclonal specifically detects GRB10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GRB10 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Grb-10, GRB10 adapter protein, GRB10 adaptor protein, GRBIR, GRB-IR, growth factor receptor-bound protein 10, Insulin receptor-binding protein Grb-IR, IRBP, KIAA0207, maternally expressed gene 1, MEG1, RSS | |
| GRB10 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS with 0.02% Sodium Azide | |
| 2887 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human GRB10. The immunogen is located within amino acids 440 - 490 of GRB10. Amino Acid Squence: EEGHAWRKRSTRMNILG | |
| Primary | |
| At least three isoforms of GRB10 are known to exist; this antibody will detect all three isoforms. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title