Learn More
GRB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10254-2-AP-20UL
 View more versions of this product
Product Code. 16856523
  • € 145.00 / 20µL
Description
GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2 - Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.
Specifications
Specifications
GRB2
Polyclonal
Unconjugated
GRB2
Adapter protein GRB2, ASH, EGFRBP GRB2, GRB2, Grb3 3, MST084, MSTP084, Protein Ash, SH2/SH3 adapter GRB2
Rabbit
Antigen Affinity Chromatography
RUO
14784, 2885, 81504
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62993, P62994, Q60631
GRB2
GRB2 Fusion Protein Ag0395
20 μL
Primary
Rat, Human, Mouse