Learn More
Ansell™ Comfort G26G Nitrile Immersion Gloves
Shop All Ansell Products
Click to view available options
Size:
6.5
7.5
8.5
10.5
Unit Size:
12 pairs
Description
- Very good abrasion resistance.
- Allows contact up to 100°C.
- Superior raised grip pattern for safety handlings.
- Open cuff with textile edge binding allows ventilation to keep hand cool.
Specifications
Specifications
| Back Material | Nitrile |
| Cuff Material | Nitrile |
| Product Type | Immersion Gloves |
| Size | 7.5 |
| Glove Cuff Color | Green |
| Glove Cuff Style | Gauntlet |
| Liner Material | Cotton |
| Model | COMFORT G26G |
| Quantity | 12 Pairs |
| Resistant To | Excellent resistance to many solvents and oil based chemicals |