missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GS 6201
GREENER_CHOICE

Product Code. 30001649
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001649

Brand: Tocris Bioscience™ 4727/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective A2B antagonist

Selective antagonist of adenosine A2B receptors (Ki values are 22, 1070, 1940 and 3280 nM for human A2B, A3, A1 and A2A receptors respectively). Attenuates allergen-, NECA- and AMP-induced airway reactivity in an allergic mouse model of asthma. Also shown to attenuate the inflammatory response during acute myocardial infarction in mice; reduces caspase-1 activity in the heart.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 752222-83-6
Molecular Formula C21H21F3N6O2

Specifications

Quantity 50 mg
Formula Weight 446.43
Purity >99%
Molecular Weight (g/mol) 446.434
Chemical Name or Material GS 6201
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.