missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 3787
GREENER_CHOICE

Product Code. 18025332
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025332 10 mg 10mg
18063922 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025332 Supplier Tocris Bioscience™ Supplier No. 3961/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PPARδ antagonist

Potent and selective peroxisome proliferator-activated receptor δ (PPARδ) antagonist (pIC50 = 6.6). Displays no measurable affinity for PPARα or PPARγ in vitro (pIC50< 5).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188591-46-0
Molecular Formula C15H12ClF3N2O3S

Specifications

Quantity 10 mg
Formula Weight 392.78
Purity >99%
Molecular Weight (g/mol) 392.78
Chemical Name or Material GSK 3787
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.