missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119150
This item is not returnable.
View return policy
Description
GSTT2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Has a sulfatase activity.
Specifications
| GSTT2 | |
| Polyclonal | |
| Unconjugated | |
| Gstt2 | |
| AI266894; Glutathione S-transferase 12; glutathione S-transferase theta 2 (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase Yrs-Yrs; glutathione S-transferase, theta 2; GST 12-12; GST class-theta-2; GSTT2; mGSTT2; Yrs | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 14872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q61133 | |
| Gstt2 | |
| Recombinant protein Glutathione S Transferase Theta 2. The antigen corresponds to amino acid range 15-216 of the target protein. | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction