missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GSTT2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 156.00 - € 441.00

Specifications

Antigen GSTT2
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17720564
GREENER_CHOICE
View Documents
Invitrogen™
PA5119150
200 μL
€ 441.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17753334
View Documents
Invitrogen™
PA5121937
20 μL
€ 156.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GSTT2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Has a sulfatase activity.
Specifications

Specifications

GSTT2
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Mouse
Gstt2
AI266894; Glutathione S-transferase 12; glutathione S-transferase theta 2 (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase Yrs-Yrs; glutathione S-transferase, theta 2; GST 12-12; GST class-theta-2; GSTT2; mGSTT2; Yrs
Gstt2
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q61133
14872
Recombinant protein Glutathione S Transferase Theta 2. The antigen corresponds to amino acid range 15-216 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.