Learn More
Invitrogen™ GTF2F2 Recombinant Rabbit Monoclonal Antibody (24GB6035)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557419
 View more versions of this product
Product Code. 30275285
  • € 198.00 / 20µL
Description
GTF2F2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GTF2F2
Recombinant Monoclonal
Unconjugated
GTF2F2
1110031C13Rik; AI326315; ATP-dependent helicase GTF2F2; BTF4; General transcription factor IIF 30 kDa subunit; general transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; GTF2F2; RAP30; TF2F2; TFIIF; TFIIF beta; TFIIF-beta; transcription factor gamma; transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30; Unknown (protein for MGC:133695)
Rabbit
Affinity chromatography
RUO
2963, 68705, 81674
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB6035
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P13984, Q01750, Q8R0A0
GTF2F2
A synthesized peptide derived from human GTF2F2 (aa 1-29).
20 μL
Primary
Human, Mouse, Rat