missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2F2 Recombinant Rabbit Monoclonal Antibody (24GB6035)
Rabbit Recombinant Monoclonal Antibody
€ 198.00 - € 564.00
Specifications
| Antigen | GTF2F2 |
|---|---|
| Clone | 24GB6035 |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30275737
|
Invitrogen™
MA557418 |
100 μL |
€ 564.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275285
|
Invitrogen™
MA557419 |
20 μL |
€ 198.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTF2F2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.Specifications
| GTF2F2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GTF2F2 | |
| 1110031C13Rik; AI326315; ATP-dependent helicase GTF2F2; BTF4; General transcription factor IIF 30 kDa subunit; general transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; GTF2F2; RAP30; TF2F2; TFIIF; TFIIF beta; TFIIF-beta; transcription factor gamma; transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30; Unknown (protein for MGC:133695) | |
| GTF2F2 | |
| Primary | |
| Affinity chromatography |
| 24GB6035 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13984, Q01750, Q8R0A0 | |
| 2963, 68705, 81674 | |
| A synthesized peptide derived from human GTF2F2 (aa 1-29). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title