Learn More
HBsAg Antibody (Hs33), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-73138AF594
 View more versions of this product
Product Code. 18742300
  • € 519.00 / 0.20mL
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HBsAg
Monoclonal
Alexa Fluor 594
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBsAg of ayw subtype.
0.2 mL
Cell Biology, Virology Bacteria and Parasites
944569
Store at 4°C in the dark.
IgG2a
Sandwich ELISA
Hs33
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Virus