missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (Hs33), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
€ 519.00
Specifications
| Antigen | HBsAg |
|---|---|
| Clone | Hs33 |
| Applications | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 594 |
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISASpecifications
| HBsAg | |
| Sandwich ELISA | |
| Alexa Fluor 594 | |
| Mouse | |
| Cell Biology, Virology Bacteria and Parasites | |
| 50mM Sodium Borate | |
| 944569 | |
| IgG2a | |
| Protein G purified |
| Hs33 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBsAg of ayw subtype. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title