missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC11 Recombinant Rabbit Monoclonal Antibody (23GB6005), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552970
This item is not returnable.
View return policy
Description
This gene encodes a class IV histone deacetylase. The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants.
Specifications
| HDAC11 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HDAC11 | |
| FLJ22237; HD11; HDAC11; Histone deacetylase 11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232232, 79885 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB6005 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91WA3, Q96DB2 | |
| HDAC11 | |
| A synthesized peptide derived from human HDAC11 (205-269AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction