missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2146-0.025ml
This item is not returnable.
View return policy
Description
HDAC7 Polyclonal specifically detects HDAC7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| HDAC7 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500 | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Predicted molecular weight: 105 kDa. Additional bands may be detected in various extract preparations. Detection of HDAC7 is specifically inhibited with the immunizing peptide. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
| HDAC7 | |
| Synthetic peptide corresponding to amino acids 357-373 of Human HDAC7 (RSEPLPPSATAPPPPGP) conjugated to KLH. The corresponding sequence is identical in mouse. | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
| 51564 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction