missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 193.00
Specifications
| Antigen | HDAC7 |
|---|---|
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
HDAC7 Polyclonal specifically detects HDAC7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| HDAC7 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| HDAC7 | |
| IgG | |
| Affinity Purified | |
| Predicted molecular weight: 105 kDa. Additional bands may be detected in various extract preparations. Detection of HDAC7 is specifically inhibited with the immunizing peptide. |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
| 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide | |
| 51564 | |
| Synthetic peptide corresponding to amino acids 357-373 of Human HDAC7 (RSEPLPPSATAPPPPGP) conjugated to KLH. The corresponding sequence is identical in mouse. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title