missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HSA98), Novus Biologicals™
Mouse Monoclonal Antibody
€ 264.00 - € 474.00
Specifications
| Antigen | Hepatocyte Specific Antigen |
|---|---|
| Clone | HSA98 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 1-5 ug/mL |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18439481
|
Novus Biologicals
NBP2-32860-0.02mg |
0.02 mg |
€ 264.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18497241
|
Novus Biologicals
NBP2-32860-0.1mg |
0.1 mg |
€ 474.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for ELISASpecifications
| Hepatocyte Specific Antigen | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| HEP-3B human hepatocellular carcinoma cells | |
| Primary | |
| Store at 4C. |
| HSA98 | |
| ELISA 1-5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Hepatocyte, HSA | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title