missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HSA98), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32860-0.02mg
This item is not returnable.
View return policy
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for ELISA
Specifications
| Hepatocyte Specific Antigen | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 1-5 ug/mL | |
| Hepatocyte, HSA | |
| HEP-3B human hepatocellular carcinoma cells | |
| 0.02 mg | |
| Cellular Markers | |
| Human | |
| Purified |
| ELISA | |
| HSA98 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction