missing translation for 'onlineSavingsMsg'
Learn More

HEPPSO, 99%, MP Biomedicals

Product Code. 11453781 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g

Product Code. 11453781

Brand: MP Biomedicals 0215123610

Please to purchase this item. Need a web account? Register with us today!

New Zwitterionic hydrogen buffer for biological research

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-78-0
Molecular Formula C9H20N2O5S
Molecular Weight (g/mol) 268.328
InChI Key GIZQLVPDAOBAFN-UHFFFAOYSA-N
Synonym heppso, 2-hydroxy-3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, n-hydroxyethyl piperazine-n'-2-hydroxypropanesulfonic acid, beta-hydroxy-4-2-hydroxyethyl piperazine-1-propanesulphonic acid, beta-hydroxy-4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl-1-piperazinyl-2-hydroxypropanesulfonic acid, 4-2-hydroxyethyl piperazine-1-2-hydroxypropane-3-sulfonic acid, 4-2-hydroxyethyl piperazine-1-2-hydroxypropane sulfonic acid free acid, 2-hydroxy-3-4-2-hydroxyethyl piperazinyl propanesulfonic acid
PubChem CID 100205
ChEBI CHEBI:32951
IUPAC Name 2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCO)CC(CS(=O)(=O)O)O

Specifications

Quantity 10 g
Assay Percent Range 99%
Formula Weight 268.3
Chemical Name or Material HEPPSO, Free Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.