missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptelidic Acid, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.25 mg
1 mg
Unit Size:
1mg
250µg
Description
- Glyceraldehyde 3-phosphate dehydrogenase (GAPDH), a key enzyme in carbohydrate metabolism, reversibly catalyzes the conversion of GAP to 1,3-bisphosphoglycerate and NAD+
- A sesquiterpene lactone produced by the fungus T. koningii that was shown to have antibiotic activity against anaerobic bacteria such as Bacteroides
- It acts as an irreversible inhibitor of GAPDH that binds to the cysteine-149 residue at the active site of the enzyme (Ki = 1.6 μM)
- Can selectively induce apoptosis in high-glycolytic cancer cells by inhibiting the generation of ATP in the glycolytic pathway
- Heptelidic acid is also a selective and competitive inhibitor of mammalian DNA polymerases β and λ as well as terminal deoxynucleotidyl transferase in family X of DNA polymerases (Kis range from 5.2-9.5 μM)
Specifications
Specifications
| Quantity | 0.25 mg |
| Color | White |
| Purity | >95 |
| Molecular Weight (g/mol) | 280.3 |
| Chemical Name or Material | Heptelidic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction