missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HER-2 / c-erbB-2 / neu / CD340 Mouse, Clone: ERBB2/2452, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2064-MSM12-P0
This item is not returnable.
View return policy
Specifications
| HER-2 / c-erbB-2 / neu / CD340 | |
| Cell Surface | |
| ERBB2/2452 | |
| Unconjugated | |
| Mouse | |
| 185kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 2064 | |
| IgG2a κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human HER-2 protein fragment (apprx. aa 311-462) | |
| SKBR-3 Cells or Breast Cancers. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction